Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Heavy Wall Recovery Flasks with Square Bead, standard taper Outer Joint


Ideal for use with rotary evaporators

Manufacturer: kimble™  6086754324

 View more versions of this product

Catalog No. K608675-4324

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Rotary evaporator
Capacity (Metric) 200mL
Neck Style Single
Diameter (Metric) Outer 76mm
Material Borosilicate Glass
Standard Taper Joint Side 24/40
Total Count 1.0
Standard Taper Joint 24/40
For Use With (Application) Square bead joints provide superior clamping to standard Joint beads
Diameter (English) Outer 2.99 in.