Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Hamilton Syringe Luer Fittings
Manufacturer: kimble™  H350310000
 View more versions of this product
Catalog No. KH35031-0000
  • / Each
Description & Specifications
Specifications
For Use With Inert miniature and syringe valves for low pressure applications
Thread Size 1/4-28 UNF
Material CTFE
Type Luer Fitting (Female)