Kimble™ Kontes™ Hamilton Syringe Luer Fittings
$58.38 - $58.38
Specifications
For Use With Inert miniature and syringe valves for low pressure applications
Material CTFE
Thread Size 1/4-28 UNF
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
KH35030-0000 Kimble™
H350300000
Luer Fitting (Male) Each for $58.38
KH35031-0000 Kimble™
H350310000
Luer Fitting (Female) Each for $58.38
Specifications Description & Specifications
Specifications
Inert miniature and syringe valves for low pressure applications
CTFE
1/4-28 UNF