Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Flasks with Three Angle Necks and standard taper Joints


Side necks are angled off center neck for greater accessibility

Manufacturer: kimble™  2955100500

 View more versions of this product

Catalog No. K295510-0500

  • / Each
 

Description & Specifications

Specifications

Bottom Shape Spherical
Capacity (Metric) 500mL
Height (English) With Closure 5.3 in.
Includes Glass plug and the patented pressure/vacuum retainer
No. of Positions 3.0
Standard Taper Joint Side 19/22
Standard Taper Joint 14/20
For Use With (Application) For heating, stirring and solvent stripping without impairing ease of liquid and solid transfer
Height (Metric) With Closure 13.5cm
Neck Style Angled
Shape Kjeldahl
Material Borosilicate Glass