Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Flasks with Three Angle Necks and standard taper Joints


Side necks are angled off center neck for greater accessibility

Manufacturer: kimble™  2955100100

 View more versions of this product

Catalog No. K295510-0100

  • / Each
 

Description & Specifications

Specifications

Bottom Shape Spherical
Capacity (Metric) 25mL
For Use With (Application) For heating, stirring and solvent stripping without impairing ease of liquid and solid transfer
Height (English) With Closure 4.7 in.
Height (Metric) With Closure 12cm
Includes Glass plug and the patented pressure/vacuum retainer
Neck Style Angled
No. of Positions 3.0
Shape Kjeldahl
Standard Taper Joint Side 19/22
Standard Taper Joint 14/20
Material Borosilicate Glass

  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements