missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Flasks with Three Angle Necks and standard taper Joints

Side necks are angled off center neck for greater accessibility

Manufacturer:  Kimble™ 6060201024

 View more versions of this product

Catalog No. K606020-1024


 

Specifications

Description

Flask, Distilling, 500mL; Side Necks standard taper No.: 24/40 Center Neck standard taper No.: 24/40

  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

Description & Specifications

Specifications

Round
102mm
Angled
24/40 Center, 24/40 side
Borosilicate Glass
4.01 in.
Standard taper outer joint
3.0
500mL