Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Distillation Flasks


Improved version of classic Schlenk tube

Manufacturer: kimble™  2131001024

 View more versions of this product

Catalog No. K213100-1024

  • / Each
 

Description & Specifications

Specifications

Capacity (Metric) 100mL
Includes Glass plug and the patented pressure/vacuum retainer
Material Borosilicate Glass
For Use With (Application) For heating, stirring and solvent stripping without impairing ease of liquid and solid transfer
Bottom Shape Spherical
Standard Taper Joint 24/40
Shape Kjeldahl
Height (Metric) With Closure 18.5cm
Height (English) With Closure 7.2 in.