Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Distillation Flasks
Improved version of classic Schlenk tube
Manufacturer: kimble™  2131000114
 View more versions of this product
Catalog No. K213100-0114
  • / Each
Description & Specifications
Specifications
Bottom Shape Spherical
For Use With (Application) For heating, stirring and solvent stripping without impairing ease of liquid and solid transfer
Capacity (Metric) 10mL
Height (English) With Closure 4 in.
Height (Metric) With Closure 10.5cm
Includes Glass plug and the patented pressure/vacuum retainer
Shape Kjeldahl
Standard Taper Joint 14/20
Material Borosilicate Glass