Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Distillation Adapters Bent with Hose Connector and Drip Tip
Bent at 105°, with hose connection and drip tip
$99.40 - $126.11
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Distillation Adapter
For Use With (Application) Distillation
Includes Hose connector 2, drip tip
View More Specs
 Alert
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Products
Catalog Number Mfr. No. Fits Tubing ID (English) Length (English) Standard Taper Joint No Price Quantity    
K205500-2440
  Accessories
kimble™
2055002440
0.37 in. 5.7 in. 24/40 Each for $126.11
K276410-0000
kimble™
2764100000
0.25 in. 4.09 in. 19/22 Each for $99.40
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Distillation Adapter
For Use With (Application) Distillation
Includes Hose connector 2, drip tip