Kimble™ Kontes™ Distillation Adapters
$84.49 - $128.44
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type 75° Connecting Distillation Adapter
For Use With (Application) Atmospheric or vacuum distillations
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Height (English) Height (Metric) Width (English) Standard Taper Joint No. Price Quantity  
Catalog Number Mfr. No. Height (English) Height (Metric) Width (English) Standard Taper Joint No. Price Quantity  
K167100-2440 Kimble™
1671002440
6.1 in. 155mm 3.93 in. 24/40 Each for $119.58
K167000-2440 Kimble™
1670002440
6.1 in. 155mm 3.93 in. 24/40 Each for $113.18
K167500-2440 Kimble™
1675002440
6.1 in. 155mm 4.13 in. 24/40 Each for $128.44
K274950-0000 Kimble™
2749500000
4.13 in. 105mm 2.16 in. 14/20 Each for $98.93
K275060-0000 Kimble™
2750600000
4.13 in. 105mm 2.95 in. 19/22 Each for $94.55
K274960-0000 Kimble™
2749600000
2.95 in. 75mm 1.96 in. 14/20 Each for $84.49
Specifications Description & Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
Borosilicate Glass
75° Connecting Distillation Adapter
Atmospheric or vacuum distillations