Please login to your online account to display your discounted pricing

Kontes™ Components for Ultra-Ware™ Filtration Cap System


Various filtration-cap system accessories

Manufacturer: kimble chase life science  9539140001

 View more versions of this product

Catalog No. K953914-0001

  • / Each
 

Description & Specifications

Specifications

No. per Case 1 Cs.
Type Support frit
For Use With (Equipment) Ultra-ware™ filtration cap system

  • Coarse-porosity glass support frit: 47mm (1.85 in.)
  • Upper screw clamp: 47mm (1.85 in.)
  • Drip tip: PTFE
  • Hose barb: PFA, 6.35mm (0.25 in.)
  • Filtration cap: GL 45