Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Claisen Adaptersdi


Manufacturer: kimble™  2737600000

 View more versions of this product

Catalog No. K2737600000

  • / Each
 

Description & Specifications

Specifications

Width (English) 2.83 in.
Standard Taper Joint No. 19/22
Height (English) 4.92 in.
Height (Metric) 125mm
Width (Metric) 72mm
Product Type Claisen Distillation Adapter
For Use With (Application) For temperature measurement at either top outer opening