Kimble™ Kontes™ Chromatography Packing Reservoir, with Joints
These heavy duty reservoirs are designed for use with 420510 and 420550 series chromatography columns with and without fritted glass supports or in combination with 420575 series flow control adapters.
Manufacturer:  Kimble™ 4205701024
 View more versions of this product
Catalog No. K4205701024
Specifications
Description
Reservoir, Packing; Kimble/Kontes; With joints; 24/40 upper joint; 24/40 lower joint; 1000mL capacity
  • Reservoirs are available in a variety of sizes and joint configurations
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Description & Specifications
Specifications
Packing Reservoir, with Joints