DWK Life Sciences Kimble™ Kontes™ Chromatography Column with Fritted Disc, PTFE Stopcock Plug and Joint
Gravity chromatography column
Supplier: DWK Life Sciences 4205500560
Description
- Medium wall tubing
- 40-60 micron pore size
Specifications
50 mm | |
420570 Series reservoir and 420575 series flow control adapters | |
1175 mL | |
Glass |
Gravity Chromatography Column | |
40 to 60 μm | |
600 mm | |
Fritted disc, 2mm bore PTFE stopcock plug and 24/40 standard taper upper joint |