Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Butyl Rubber Stoppers


Manufacturer: kimble™  7382713

 View more versions of this product

Catalog No. 03-340-71Z

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Size 13
Product Type Stopper, Plug style
Stopper Number 13mm
For Use With (Equipment) Fits GPI aluminum seal finish vials
For Use With (Application) Low temperature applications
Material Rubber
Color Gray