Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Butyl Rubber Stoppers


Manufacturer: kimble™  7382711

 View more versions of this product

Catalog No. 03-340-71Y

  • / Case of 1000 Each
 

Description & Specifications

Specifications

For Use With (Application) Low temperature applications
For Use With (Equipment) Fits GPI aluminum seal finish vials
Size 11
Stopper Number 11mm
Color Gray
Material Rubber
Product Type Stopper, Plug style