missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Brand 3mm NMR Tubes

Each tube meets rigid specifications for wall thickness, variation and camber so as to assure precise measurements and reproducibility.

Manufacturer:  Kimble™ 8978350008

 View more versions of this product

Catalog No. K897835-0008


 

Specifications Description & Specifications

Specifications

Borosilicate Glass
Compression-fit
±0.001 in.
3mm
2.4mm
500MHz
Cap
Cylindrical
0.28mm
Polyethylene
203.2mm
0.094 in.
±0.0006 in.
0.11 in.
Research applications include analysis of polymer structures and organic solutions
8 in.
0.011 in.