Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Brand 10mm NMR Sample Tubes


Precision tubes

Manufacturer: kimble™  8973200008

 View more versions of this product

Catalog No. K8973200008

  • / Each
 

Description & Specifications

Specifications

Length (Metric) 20cm
Field Strength 60MHz
Material Borosilicate Glass