missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Kontes™ Bevel-Seal Vacuum Inlet Adapters

For use with an O.D. from 2 to 17mm

Supplier:  DWK Life Sciences 1797501129

 View more versions of this product

Catalog No. K179750-1129



Description

Description

  • Includes one Viton O-ring and one cap

Specifications

Specifications

USP Type I, ASTM E438, Type I, Class A
0.37 in.
Compression cap, viton O-ring, hose connection No. 2
Vacuum Inlet Adapter
20-415 GPI
For plain thermometers, pipets, syringes or other small items with an O.D. from 2 to 11mm
9.39mm
Borosilicate Glass
29/42
SDS
Documents

Documents

Promotions

Promotions