Kimble™ Kontes™ Bevel-Seal Vacuum Inlet Adapters
For use with an O.D. from 2 to 17mm
Manufacturer:  Kimble™ 1797500824
 View more versions of this product
Catalog No. K179750-0824
Specifications
Description
Adapters, Inlet; Kimble/Kontes; Vacuum; Bevel-Seal; For use with an O.D. from 2mm to 17mm; Includes one Viton o-ring and one cap; 24/40 joint; 5-8mm accom. range; 1/4 in. nom.; 108 o-ring; 15-415 modified GPI thread; #2 hose connection
  • Includes one Viton O-ring and one cap
Description & Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
9.39mm
One Viton O-ring and one cap, hose connection No. 2
Borosilicate Glass
24/40
0.37 in.
For plain thermometers, pipets, syringes or other small items with an O.D. from 2 to 11mm
15-415 GPI
Vacuum Inlet Adapter