Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Bevel-Seal Vacuum Inlet Adapters
For use with an O.D. from 2 to 17mm
$72.62 - $114.38
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Vacuum Inlet Adapter
For Use With (Application) For plain thermometers, pipets, syringes or other small items with an O.D. from 2 to 11mm
View More Specs
Products
Catalog Number Mfr. No. Fits Tubing ID (English) Fits Tubing ID (Metric) Thread Size Standard Taper Joint No. Price Quantity    
K179750-0829
kimble™
1797500829
0.37 in. 9.39mm 15-415 GPI 29/42 Each for $95.20
K179750-0524
kimble™
1797500524
0.37 in. 9.39mm 13-425 GPI 24/40, 29/42 Each for $73.56
K179750-1119
kimble™
1797501119
0.25 in. 6.35mm 20-415 GPI 19/22 Each for $107.44
K179750-0814
kimble™
1797500814
0.25 in. 6.35mm 15-415 GPI 14/20, 24/40 Each for $73.56
K179750-0824
kimble™
1797500824
0.37 in. 9.39mm 15-415 GPI 24/40 Each for $73.56
K179750-1129
kimble™
1797501129
0.37 in. 9.39mm 20-415 GPI 29/42 Each for $114.38
K179750-2129
kimble™
1797502129
0.37 in. 9.39mm 13-425 GPI 29/42 Each for $72.62
K179750-0514
kimble™
1797500514
0.25 in. 6.35mm 13-425 GPI 14/20 Each for $73.56
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Vacuum Inlet Adapter
For Use With (Application) For plain thermometers, pipets, syringes or other small items with an O.D. from 2 to 11mm
  • Includes one Viton O-ring and one cap