Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Bevel-Seal™ Inlet Adapter
For use with pipets, syringes, or other small items with outer diameters from 0.06 to 0.62 in. (2 to 17mm)
Manufacturer: kimble™  1797000814
 View more versions of this product
Catalog No. K179700-0814
  • / Case of 20 Each
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) For plain thermometers, pipets, syringes or other small items with an O.D. from 2 to 17mm
Includes Cap and Viton™ O-ring
Thread Size 13-415
Material Borosilicate Glass
Product Type Bevel-Seal™ Inlet Adapter
Standard Taper Joint No. 14/20