Kimble™ Kontes™ Bevel-Seal™ Inlet Adapter
For use with pipets, syringes, or other small items with outer diameters from 0.06 to 0.62 in. (2 to 17mm)
Manufacturer:  Kimble™ 1797000529
 View more versions of this product
Catalog No. K179700-0529
Specifications Description & Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
Borosilicate Glass
29/42
Bevel-Seal™ Inlet Adapter
Cap and Viton™ O-ring
For plain thermometers, pipets, syringes or other small items with an O.D. from 2 to 17mm
13-415