missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Kontes™ Airless-Ware PTFE Plug Gas Inlet Adapter
Inlet Adapter for Vacuum and AIRLESS-WARE™, with a choice of 14/20 or 24/40 standard taper joints.
Supplier: DWK Life Sciences 2113302440
Description
- Inner Standard Taper joints
- 211330 series has a PTFE plug
- Manufactured from 33 expansion, low extractable borosilicate glass
- Conforms to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
USP Type I, ASTM E438, Type I, Class A | |
135mm | |
Borosilicate Glass | |
24/40 Outer | |
60mm |
5.31 in. | |
Patented 809000 pressure/vacuum retainer | |
Inlet Adapter | |
2.36 in. |