Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Airless-Ware Glass Plug Gas Inlet Adapter


Manufacturer: kimble™  2113002440

 View more versions of this product

Catalog No. K211300-2440

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Height (English) 5.31 in.
Height (Metric) 135mm
Includes Patented 809000 pressure/vacuum retainer
Width (English) 2.36 in.
Width (Metric) 60mm
Material Borosilicate Glass
Product Type Inlet Adapter
Standard Taper Joint No. 24/40 Inner