Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Adapters, 105 Degrees
$52.40 - $106.24
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Distillation Bent Adapter
View More Specs
Products
Catalog Number Mfr. No. Height (English) Height (Metric) Includes Standard Taper Joint No. Price Quantity    
K273100-0000
kimble™
2731000000
2.55 in. 65mm Tapered drip tube 14/20 Each for $52.40
K273200-1922
kimble™
2732001922
2.75 in. 70mm Drip tip 19/22 Each for $81.85
K276400-0000
kimble™
2764000000
4 in. 104mm Vacuum take-off, a hose connector and a drip tip 14/20 Each for $102.27
K273200-0000
kimble™
2732000000
2.75 in. 70mm Drip tip 14/20 Each for $86.85
K276405-0000
kimble™
2764050000
3.93 in. 100mm Vacuum take-off, a hose connector and a drip tip 14/20 Each for $83.67
K157000-2440
kimble™
1570002440
4.52 in. 115mm Tapered drip tube 24/40 Each for $67.56
K157500-2440
kimble™
1575002440
5.9 in. 150mm Outer joint at top 29/42 Each for $106.24
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Product Type Distillation Bent Adapter