Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Adapters, 105 Degrees


Manufacturer: kimble™  2764000000

 View more versions of this product

Catalog No. K276400-0000

  • / Each
 

Description & Specifications

Specifications

Standard Taper Joint No. 14/20
Height (English) 4 in.
Height (Metric) 104mm
Fits Tubing ID (English) 0.25 in.
Fits Tubing ID (Metric) 6.35mm
Product Type Distillation Bent Adapter
Includes Vacuum take-off, a hose connector and a drip tip
Material Borosilicate Glass
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A