Kimble™ Kontes™ Adapters, 105 Degrees
Manufacturer:  Kimble™ 2732001922
 View more versions of this product
Catalog No. K273200-1922
Specifications Description & Specifications
Specifications
Distillation Bent Adapter
2.75 in.
USP Type I, ASTM E438, Type I, Class A
55mm
19/22
Borosilicate Glass
2.16 in.
70mm
Drip tip