missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kojic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205220050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Kojic acid | |
151.0°C to 156.0°C | |
99% | |
C6H6O4 | |
5 g | |
15,5365 | |
Solubility in water: soluble. Other solubilities: freely soluble in acetone and ethanol;,sparingly soluble in diethyl ether,ethyl acetate,,chloroform and pyridine;,insoluble in benezne | |
OCC1=CC(=O)C(O)=CO1 | |
142.11 | |
CHEBI:43572 | |
99% |
501-30-4 | |
Authentic | |
Glass bottle | |
MFCD00006580 | |
18,II,57 | |
kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52 | |
BEJNERDRQOWKJM-UHFFFAOYSA-N | |
5-hydroxy-2-(hydroxymethyl)pyran-4-one | |
3840 | |
142.11 | |
Crystalline Powder |
Chemical Identifiers
501-30-4 | |
142.11 | |
BEJNERDRQOWKJM-UHFFFAOYSA-N | |
3840 | |
5-hydroxy-2-(hydroxymethyl)pyran-4-one |
C6H6O4 | |
MFCD00006580 | |
kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52 | |
CHEBI:43572 | |
OCC1=CC(=O)C(O)=CO1 |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 207-922-4
RUO – Research Use Only