Koehler™ Instrument LPG Copper Strip Corrosion Test Cylinder and Accessories
Supplier:  Koehler™ Instrument K40200
 View more versions of this product
Catalog No. KL-K40200
Description
Description
Specifications
Specifications
3.2mm Hole per ASTM specifications
Each
For LPG
Copper Strip
Stainless Steel
ASTM D1838, GPA 2140, ISO 6251, CE