Please login to your online account to display your discounted pricing
Koehler™ Instrument Copper Strip Tarnish Test Accessories
Various accessories associated with Copper Strip Tarnish Test
Manufacturer: koehler inst co  KOEK25100
 View more versions of this product
Catalog No. KL-K25100
  • / Each
Description & Specifications
Specifications
Certifications/Compliance ASTM
For Use With (Application) Provide colored reproductions of tarnished strips encased in a plastic plaque
For Use With (Equipment) Koehler™ K25339 copper strip tarnish test apparatus
Product Type Copper Corrosion Standard Strip