Carestream Health BioMax™ XAR™ Film
Suitable for use with all radioisotopes and chemiluminescent probes
Manufacturer: Carestream Health 1651512
Description
- Coated on both sides of a clear base, making it ideal for both direct autoradiography and exposures with intensifying screens Utilizes an improved formulation that eliminates the need for interleaving paper while maintaining the same performance as the original X-Omat™ AR (XAR) film Suitable for manual and automated processing
Order Info
Formats include: Bulk-packed film; ReadyPacked film (with each sheet of film individually wrapped in a protective folder for easy handling)
Specifications
X-OMAT AR-5 Film | |
35 cm | |
43 cm | |
For all radioisotopes and chemiluminescent probes, ideal for both direct autoradiography and exposures with intensifying screens utilizes an improved formulation |
14 in. | |
17 in. | |
14 x 17 in. (35 x 43 cm) | |
50 Pack, Bulk Pack |