Carestream Health BioMax™ TranScreen™ HE Autoradiography Intensifying Screens
Innovative intensifying screen developed specifically for detection of higher energy radioisotopes such as 32P and 125I
Manufacturer:  Carestream Health 8563959
 View more versions of this product
Catalog No. 05-728-63
Description
Description
Equals the sensitivity of the BioMax™ MS screen with improved handling features and publication-quality images
Order Info
Available in two formats: Intensifying screen only; or with BioMax cassette (see 05-728-61).
Specifications
Specifications
BioMax MS film
8 in.
10 in.
8 x 10 in. (203 x 254 mm)
Intensifying Screen
203 mm
254 mm
Detection of higher energy radioisotopes such as 32P and 125I
Documents