Carestream Health BioMax™ TranScreen™ HE Autoradiography Intensifying Screens
Innovative intensifying screen developed specifically for detection of higher energy radioisotopes such as 32P and 125I
Manufacturer: Carestream Health 8811457
Description
Equals the sensitivity of the BioMax™ MS screen with improved handling features and publication-quality images
Order Info
Available in two formats: Intensifying screen only; or with BioMax cassette (see 05-728-61).
Specifications
BioMax MS film | |
14 in. | |
17 in. | |
14 x 17 in. (355.6 x 431.8 mm) |
Intensifying Screen | |
355.6 mm | |
431.8 mm | |
Detection of higher energy radioisotopes such as 32P and 125I |