Carestream Health BioMax™ MS (Maximum Sensitivity) Film
Ideal for use with radioisotopes when detection of low-intensity samples or reduction of exposure times is required
Manufacturer: Carestream Health 8294985
Description
Use for detection of Northern, Southern and dot blotting applications. Suitable for manual and automated processing.
- Coated on both sides of a clear base with an advanced technology “T-grain” emulsion
- When used in conjunction with BioMax MS or TranScreen™ HE intensifying screens, film provides the fastest and most sensitive system for the detection of 32P and 125I radioisotopes (4 to 8 times faster than any other filter/screen combination)
- When combined with TranScreen LE intensifying screen, film provides similar speeds to BioMax XAR™ with a calcium tungstate screen, but with significantly greater resolution
- When used with TranScreen LE, film offers the fastest system for the detection of 35S, 14C, 45Ca, 33P, 59Fe, and even 3H radioisotopes (≥5 times faster)
May be used with other intensifying screens; however, maximum sensitivity is achieved using BioMax screens.
Order Info
Choose noninterleaved, bulk-packed film or ReadyPacked film (each sheet individually wrapped in a protective folder).
Specifications
Maximum Sensitivity Film | |
8 in. | |
10 in. | |
8 x 10 in. (20 x 25 cm) | |
50 Pack |
Bulk Packed | |
20 cm | |
25 cm | |
Ideal for use with radioisotopes, detection of northern, southern and dot blotting applications, suitable for manual and automated processing |