Carestream Health BioMax™ MS (Maximum Sensitivity) Film
Ideal for use with radioisotopes when detection of low-intensity samples or reduction of exposure times is required
Manufacturer:  Carestream Health 1435726
 View more versions of this product
Catalog No. 05-728-2
Description
Description
Use for detection of Northern, Southern and dot blotting applications. Suitable for manual and automated processing.
  • Coated on both sides of a clear base with an advanced technology “T-grain” emulsion
  • When used in conjunction with BioMax MS or TranScreen™ HE intensifying screens, film provides the fastest and most sensitive system for the detection of 32P and 125I radioisotopes (4 to 8 times faster than any other filter/screen combination)
  • When combined with TranScreen LE intensifying screen, film provides similar speeds to BioMax XAR™ with a calcium tungstate screen, but with significantly greater resolution
  • When used with TranScreen LE, film offers the fastest system for the detection of 35S, 14C, 45Ca, 33P, 59Fe, and even 3H radioisotopes (≥5 times faster)
May be used with other intensifying screens; however, maximum sensitivity is achieved using BioMax screens.
Order Info
Choose noninterleaved, bulk-packed film or ReadyPacked film (each sheet individually wrapped in a protective folder).
Specifications
Specifications
17 in.
Ideal for use with radioisotopes, detection of northern, southern and dot blotting applications, suitable for manual and automated processing
43cm
Maximum Sensitivity Film
Bulk Packed
14 x 17 in. (35 x 43cm)
35cm
14 in.
50 Pack
Documents