Carestream Health BioMax™ MS (Maximum Sensitivity) Film
Ideal for use with radioisotopes when detection of low-intensity samples or reduction of exposure times is required
Manufacturer:  Carestream Health 8222648
 View more versions of this product
Catalog No. 05-728-4
Description
Description
Use for detection of Northern, Southern and dot blotting applications. Suitable for manual and automated processing.
  • Coated on both sides of a clear base with an advanced technology “T-grain” emulsion
  • When used in conjunction with BioMax MS or TranScreen™ HE intensifying screens, film provides the fastest and most sensitive system for the detection of 32P and 125I radioisotopes (4 to 8 times faster than any other filter/screen combination)
  • When combined with TranScreen LE intensifying screen, film provides similar speeds to BioMax XAR™ with a calcium tungstate screen, but with significantly greater resolution
  • When used with TranScreen LE, film offers the fastest system for the detection of 35S, 14C, 45Ca, 33P, 59Fe, and even 3H radioisotopes (≥5 times faster)
May be used with other intensifying screens; however, maximum sensitivity is achieved using BioMax screens.
Order Info
Choose noninterleaved, bulk-packed film or ReadyPacked film (each sheet individually wrapped in a protective folder).
Specifications
Specifications
7 x 9.5 in. (18 x 24 cm)
Bulk Packed
18 cm
24 cm
50 Pack
Maximum Sensitivity Film
7 in.
9.5 in.
Ideal for use with radioisotopes, detection of northern, southern and dot blotting applications, suitable for manual and automated processing