Carestream Health BioMax™ MS Autoradiography Intensifying Screens
Conventional format intensifying screens for maximum sensitivity in detection of higher energy radioisotopes such as 32P and 125I
Manufacturer:  Carestream Health 8518706
 View more versions of this product
Catalog No. 05-728-23
Description
Description
Four to eight times faster than other autoradiography screens
Available in two formats
  • Intensifying screen only
  • With BioMax cassette (see 05-728-17 and 05-728-16)
Specifications
Specifications
8 x 10 in. (203 x 254 mm)
BioMax MS film
203.2 mm
8 in.
10 in.
Detection of 32 P and 125 I labeled samples
BioMax Cassette
254 mm
BioMax MS Intensifying Screen
Documents