Carestream Health BioMax™ MS Autoradiography Intensifying Screens
Conventional format intensifying screens for maximum sensitivity in detection of higher energy radioisotopes such as 32P and 125I
Manufacturer:  Carestream Health 8981706
 View more versions of this product
Catalog No. 05-728-22
Description
Description
Four to eight times faster than other autoradiography screens
Available in two formats
  • Intensifying screen only
  • With BioMax cassette (see 05-728-17 and 05-728-16)
Specifications
Specifications
BioMax MS film
14 in.
17 in.
14 x 17 in. (355.6 x 431.8 mm)
Detection of 32 P and 125 I labeled samples
BioMax MS Intensifying Screen
355.6 mm
431.8 mm
BioMax Cassette