Carestream Health BioMax™ MR (Maximum Resolution) Film
Recommended for direct autoradiography of weak and medium energy β-radioisotopes, such as 35S,33P, and 14C
$898.38 - $5,244.26
Specifications
For Use With (Application) Provides maximum resolution and sensitivity for detection of 35 S, 33 P, and 14 C chemiluminescent labeled samples, ideal for the detection of sequencing gels coated on only one side of a clear base Utilizes patented “T-grain” technology
View More Specs
Products 8
Catalog Number Mfr. No. Dimensions (LxW) Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Dimensions (LxW) Product Type Quantity Price Quantity  
05-728-25
View Documents
Carestream Health
8715187
14 x 17 in. (35.56 x 43 cm) BioMax MR-1 Film: Noninterleaved 50 Pack, Bulk Pack
Pack of 50 Each for $3,598.46
Case of 5 Pack for $17,992.30
05-728-27
View Documents
Carestream Health
8736936
7 x 9.5 in. (17.78 x 24 cm) BioMax MR-1 Film: Noninterleaved 50 Pack, Bulk Pack
Pack of 50 Each for $1,750.39
Case of 5 Pack for $8,751.95
05-728-26
View Documents
Carestream Health
8941114
5 x 7 in. (12.7 x 18 cm) BioMax MR-1 Film: Noninterleaved 50 Pack, Bulk Pack
Pack of 50 Each for $898.38
Case of 5 Pack for $4,491.90
05-728-29
View Documents
Carestream Health
8912560
9.5 x 12 in. (24.13 x 30cm) BioMax MR-1 Film: Noninterleaved 50 Pack, Bulk Pack
Pack of 50 Each for $2,896.45
Case of 5 Pack for $14,482.25
05-728-31
View Documents
Carestream Health
8952855
8 x 10 in. (20.32 x 25 cm) BioMax MR-2 Film: ReadyPacked 50 Pack, Individually wrapped
Pack of 50 Each for $2,977.71
Case of 5 Pack for $14,888.55
05-728-33
View Documents
Carestream Health
8572786
5 x 7 in. (12.7 x 18 cm) BioMax MR-2 Film: ReadyPacked 50 Pack, Individually wrapped
Pack of 50 Each for $2,244.03
Case of 5 Pack for $11,220.15
05-728-32
View Documents
Carestream Health
8567232
14 x 17 in. (35.56 x 43 cm) BioMax MR-2 Film: ReadyPacked 50 Pack, Individually wrapped
Pack of 50 Each for $5,244.26
Case of 2 Pack for $10,488.52
05-728-24
View Documents
Carestream Health
8701302
8 x 10 in. (20 x 25 cm) BioMax MR-1 Film: Noninterleaved 50 Pack, Bulk Pack
Pack of 50 Each for $2,186.00
Case of 5 Pack for $10,930.00
Description
Description
  • Ideal for the detection of sequencing gels Coated on only one side of a clear base Utilizes patented “T-grain” technology, which provides a two- to four-fold increase in sensitivity for direct autoradiography when compared to other films Improves detection of low-intensity bands Provides enhanced resolution and exceptional clarity
  • Also provides excellent resolution for 32P and 125I, although it offers lower sensitivity than BioMax MS film Suitable for manual and automated processing
Order Info
Available in either Bulk-packed film or ReadyPacked film (with each sheet of film individually wrapped in a protective folder).
Specifications
Specifications
Provides maximum resolution and sensitivity for detection of 35 S, 33 P, and 14 C chemiluminescent labeled samples, ideal for the detection of sequencing gels coated on only one side of a clear base Utilizes patented “T-grain” technology