Carestream Health BioMax™ MR (Maximum Resolution) Film
Recommended for direct autoradiography of weak and medium energy β-radioisotopes, such as 35S,33P, and 14C
Manufacturer:  Carestream Health 8736936
 View more versions of this product
Catalog No. 05-728-27
Description
Description
  • Ideal for the detection of sequencing gels Coated on only one side of a clear base Utilizes patented “T-grain” technology, which provides a two- to four-fold increase in sensitivity for direct autoradiography when compared to other films Improves detection of low-intensity bands Provides enhanced resolution and exceptional clarity
  • Also provides excellent resolution for 32P and 125I, although it offers lower sensitivity than BioMax MS film Suitable for manual and automated processing
Order Info
Available in either Bulk-packed film or ReadyPacked film (with each sheet of film individually wrapped in a protective folder).
Specifications
Specifications
BioMax MR-1 Film: Noninterleaved
17.78 cm
24 cm
Provides maximum resolution and sensitivity for detection of 35 S, 33 P, and 14 C chemiluminescent labeled samples, ideal for the detection of sequencing gels coated on only one side of a clear base Utilizes patented “T-grain” technology
7 in.
9.5 in.
7 x 9.5 in. (17.78 x 24 cm)
50 Pack, Bulk Pack