Carestream Health BioMax™ MR (Maximum Resolution) Film
Recommended for direct autoradiography of weak and medium energy β-radioisotopes, such as 35S,33P, and 14C
Manufacturer: Carestream Health 8567232
Description
- Ideal for the detection of sequencing gels Coated on only one side of a clear base Utilizes patented “T-grain” technology, which provides a two- to four-fold increase in sensitivity for direct autoradiography when compared to other films Improves detection of low-intensity bands Provides enhanced resolution and exceptional clarity
- Also provides excellent resolution for 32P and 125I, although it offers lower sensitivity than BioMax MS film Suitable for manual and automated processing
Order Info
Available in either Bulk-packed film or ReadyPacked film (with each sheet of film individually wrapped in a protective folder).
Specifications
BioMax MR-2 Film: ReadyPacked | |
35.56 cm | |
43 cm | |
Provides maximum resolution and sensitivity for detection of 35 S, 33 P, and 14 C chemiluminescent labeled samples, ideal for the detection of sequencing gels coated on only one side of a clear base Utilizes patented “T-grain” technology |
14 in. | |
17 in. | |
14 x 17 in. (35.56 x 43 cm) | |
50 Pack, Individually wrapped |