Carestream Health BioMax™ Light Film
Developed specifically for the detection of chemiluminescent probes
Manufacturer:  Carestream Health 8194540
 View more versions of this product
Catalog No. 05-728-48
Description
Description
  • Produces high signal-to-noise ratio
  • Coated on both sides of a clear base
  • Can be used for direct autoradiography and intensifying screen exposures with commonly used isotopes, but requires longer exposure times than BioMax MS, and BioMax XAR™ film
  • Suitable for manual and automated processing
Kodak autoradiography and electron micrography films should be handled under proper safelight conditions using the Kodak Dark Room Illuminator (Mfr. No. 1132752 ). This self-contained unit is matched for use with all autoradiography and electron micrography films.Use of other safelights with improper filters or improper wattage bulbs will result in increased background fog on the exposed film and poorer images.
Order Info
Formats include: Bulk-packaged film; ReadyPacked film (with each sheet of film individually wrapped in a protective folder).
Specifications
Specifications
BioMax Light-1 Film: Noninterleaved
7 x 9.5 in.
For manual and automated processing
50 Sheets/Pk., Bulk Pack