Carestream Health BioMax™ Light Film
Developed specifically for the detection of chemiluminescent probes
$1,284.81 - $2,699.52
Specifications
For Use With (Application) For manual and automated processing
View More Specs
Products 6
Catalog Number Mfr. No. Dimensions (L x W) Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Product Type Quantity Price Quantity  
05-728-47 View Documents Carestream Health
8689358
5 x 7 in. BioMax Light-1 Film: Noninterleaved 50 Sheets/Pk., Bulk Pack Pack of 50 Each for $1,284.81
Case of 5 Pack for $6,424.05
05-728-48 View Documents Carestream Health
8194540
7 x 9.5 in. BioMax Light-1 Film: Noninterleaved 50 Sheets/Pk., Bulk Pack Pack of 50 Each for $1,660.00
Case of 5 Pack for $8,300.00
05-728-49 View Documents Carestream Health
1538966
9.5 x 12 in. BioMax Light-1 Film: Noninterleaved 50 Sheets/Pk., Bulk Pack Pack of 50 Each for $2,217.45
Case of 5 Pack for $11,087.25
05-728-53 View Documents Carestream Health
1917012
5 x 7 in. BioMax Light-2 Film: ReadyPacked 50 Sheets/Pk., Individually wrapped Pack of 50 Each for $2,033.97
Case of 5 Pack for $10,169.85
05-728-52 View Documents Carestream Health
8761520
8 x 10 in. BioMax Light-2 Film: ReadyPacked 50 Sheets/Pk., Individually wrapped Pack of 50 Each for $2,699.52
Case of 5 Pack for $13,497.60
05-728-55 View Documents Carestream Health
1788207
8 x 10 in. BioMax Light-1 Film: Noninterleaved 50 Sheets/Pk., Bulk Pack Pack of 50 Each for $1,624.39
Case of 5 Pack for $8,121.95
Description
Description
  • Produces high signal-to-noise ratio
  • Coated on both sides of a clear base
  • Can be used for direct autoradiography and intensifying screen exposures with commonly used isotopes, but requires longer exposure times than BioMax MS, and BioMax XAR™ film
  • Suitable for manual and automated processing
Kodak autoradiography and electron micrography films should be handled under proper safelight conditions using the Kodak Dark Room Illuminator (Mfr. No. 1132752 ). This self-contained unit is matched for use with all autoradiography and electron micrography films.Use of other safelights with improper filters or improper wattage bulbs will result in increased background fog on the exposed film and poorer images.
Order Info
Formats include: Bulk-packaged film; ReadyPacked film (with each sheet of film individually wrapped in a protective folder).
Specifications
Specifications
For manual and automated processing