Please login to your online account to display your discounted pricing

Ko 143, Tocris

Potent and selective BCRP inhibitor

$146.74 - $316.20

Chemical Identifiers

Chemical Name or Material Ko 143
Assay Percent Range >99%
Molecular Formula C26H35N3O5
Formula Weight 469.57
Synonym (3S,6S,12aS)-1,2,3,4,6,7,12,12a-Octahydro-9-methoxy-6-(2-methylpropyl)-1,4-dioxopyrazino[1',2':1,6]pyrido[3,4-b]indole-3-propanoic acid 1,1-dimethylethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

32411

 
r & d systems
32411
1mg Each for $146.74

324110

 
r & d systems
324110
10mg Each for $316.20
Description & Specifications

Specifications

Formula Weight 469.57
Quantity 1mg
Chemical Name or Material Ko 143
CAS 461054-93-3
Synonym (3S,6S,12aS)-1,2,3,4,6,7,12,12a-Octahydro-9-methoxy-6-(2-methylpropyl)-1,4-dioxopyrazino[1',2':1,6]pyrido[3,4-b]indole-3-propanoic acid 1,1-dimethylethyl ester
Molecular Formula C26H35N3O5
Assay Percent Range >99%
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C