Please login to your online account to display your discounted pricing
KNF LIQUIPORT™ NF Series Diaphragm Liquid Pumps Standard, PTFE Head
For convenient transfer and dosing of neutral or corrosive liquids
$5,958.20 - $8,291.73
Specifications
Certifications/Compliance CE, EN 61010-1, EN 61326-1
Certifications/Compliance CE, EN 61010-1, EN 61326-1
Control Manual
Electrical Requirements 100/230V 50/60Hz
Hertz 50/60Hz
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Flow Rate Model Pressure Price Quantity    
13-880-42
knf neuberger inc
NF100FT.18S
Each for $5,958.20
13-880-45
knf neuberger inc
NF1.100FT.18S
Each for $6,471.58
13-880-48
knf neuberger inc
NF300FT.18S
Each for $7,583.18
13-880-51
knf neuberger inc
NF1.300FT.18S
Each for $8,291.73
Description & Specifications
Specifications
Certifications/Compliance CE, EN 61010-1, EN 61326-1
Certifications/Compliance CE, EN 61010-1, EN 61326-1
Control Manual
Electrical Requirements 100/230V 50/60Hz
Hertz 50/60Hz
Includes Liquiport™ Laboratory pump, Mains plug, Operating manual
IP Rating 65
Max. Temperature (English) 104°F
Max. Temperature (Metric) 40°C
Motor Speed 3000rpm
Noise Level <40dBA
Pumps are self-priming, maintenance-free and will operate dry indefinitely without damage. Select the best pump based on the desired flowrate, maximum outlet pressure and the wetted materials.
  • Premium corrosion resistance
  • Manual front-panel flowrate control
  • Pressure limit protection
  • No metal wetted parts to corrode
  • Reliable splash-proof housing
  • All models feature PTFE diaphragms and FFPM valves
Three considerations for any liquid diaphragm application:
  • The inlet of pump should not be lower than the level of liquid being transferred
  • An inlet filter is required if media contains particulates or fibers larger than 50 microns
  • Pulsation varies between specific pump models
For 100/230V 50/60Hz.