Please login to your online account to display your discounted pricing

KNF LABOPORT™ Chemically Resistant Dry Vacuum Pumps: 10L/min. Flowrate

Quiet, reliable operation without the hazards associated with handling messy pump oils or contaminated wastewater generated by aspirators

$3,033.58 - $4,047.15

Specifications

Electrical Requirements 115V 60Hz
View More Specs
Products
Catalog Number Mfr. No. Model Price Quantity    

13-878-25

 
knf neuberger inc
N8103FTP
UN810.3FTP Each for $4,047.15

13-878-40

 
knf neuberger inc
N810FTP115
UN810FTP Each for $3,033.58
Description & Specifications

Specifications

Electrical Requirements 115V 60Hz

Handle corrosive and aggressive vapors without damage.

  • Oil-free
  • End vacuums to 1.5 torr

Feature solid PTFE heads and diaphragms to protect against corrosives and aggressive vapors. Multiport Kalrez™ valves withstand the ingestion of condensed vapors without the risk of damaging the pump or degrading performance.

Recommended for:

  • rotary evaporators (13-878-27, -29, -35 and 13-880-430)
  • vacuum concentrators (13-878-29, -35 and 13-880-430)
  • gel dryers (13-878-27 and -29)
  • vacuum ovens (13-878-27, -29, -35, and 13-880-430)
  • general vacuum applications (13-878-40, -42, -44, -46, 13-880-430 and -431)

For 115V 60Hz; 230V models are also available. For information contact your Fisher Scientific™ Customer Service Representative

1For 230V 50Hz models, contact your Fisher Scientific™ Customer Service Representative.