Please login to your online account to display your discounted pricing

KM 11060, Tocris

Corrects F508del-CFTR trafficking

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material KM 11060
Assay Percent Range >98%
Molecular Formula C19H17Cl2N3O2S
Formula Weight 422.33
Synonym 7-Chloro-4-[4-[4-chlorophenyl)sulfonyl]-1-piperazinyl]quinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

314350

 
r & d systems
RDS314350
50mg Each for $812.12

314310

 
r & d systems
RDS314310
10mg Each for $192.67
Description & Specifications

Specifications

Formula Weight 422.33
Quantity 50mg
Chemical Name or Material KM 11060
Assay Percent Range >98%
Molecular Formula C19H17Cl2N3O2S
CAS 774549-97-2
Synonym 7-Chloro-4-[4-[4-chlorophenyl)sulfonyl]-1-piperazinyl]quinoline
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature