missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

KLRG1, eFluor 506, clone: 2F1, eBioscience™

Syrian Hamster Monoclonal Antibody

Manufacturer:  Life Technologies LS69589382

 View more versions of this product

Catalog No. 501124976



Description

Description

Killer cell lectin-like receptor G1 (KLRG1; formerly known as mouse MAFA or 2F1-Ag), is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1.

Description: This 2F1 monoclonal antibody reacts with the mouse Killer cell Lectin-like Receptor G1 (KLRG1), also known as Mast cell Function-associated Antigen (MAFA). KLRG1 is a homodimer of glycosylated 30-38kDa subunits and contains a cytoplasmic motif similar to the immunoreceptor tyrosine-based inhibitory motif (ITIM). Rat MAFA was identified as an antigen specific to rat mast cells; however, the expression of mouse KLRG1/MAFA using 2F1 has not been detected on the surface of mouse mast cell lines, bone marrow-derived mast cells, or peritoneal mast cells. This antigen is expressed on approximately one-third of mouse NK cells and a subset of T cells. MHC class I molecules regulate KLRG1 via interactions with class I-specific inhibitory Ly49 molecules and SHP-1 signaling. Although KLRG1 and Ly49 are both lectin-like inhibitory receptors that are regulated by class I MHC expression, the effects of this on cell surface expression of these molecules are opposing, and the underlying regulatory mechanisms distinct.Applications Reported: This 2F1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2F1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor™ 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

KLRG1
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O88713
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, MAFA-like receptor, killer cell lectin-like receptor subfamily G member 1, mast cell function-associated antigen (ITIM-containing)
Syrian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
2F1
eFluor 506
Klrg1
Liquid
Klrg1
IgG
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
50928
Documents

For Research Use Only.