BUCHI KjelMaster K-375/KjelSampler 376 System
An automatic distillation unit designed for convenient nitrogen and protein analysis. The reduced need for user interaction, and intelligent software saves time and costs.
Supplier: BUCHI 113751700
Description
- Compliant and flexible plug-and-measure titration concept (colorimetric, potentiometric)
- Comprises a steam generator, automatic aspiration of residues, glass splash protector, and integrated titration
- Fully automated dilution of the sample, alkali addition, receiver dispensing, potentiometric titration, and calculation/reporting
- Safe operation, user protection and guided routine operation
- Simple LIMS integration and traceable data recording
Specifications
220/240 V, 50/60 Hz | |
Color Touchscreen (8.4 in.) | |
15 A | |
1 Sample |
220/240 V | |
Nitrogen and Protein Analysis | |
Automatic Distillation Unit |